Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levofloxacin N-oxide | 117678-38-3 | C18H20FN3O5 | 10MG
SDP

Supplier:  Medchemexpress LLC HY13378710MG

Encompass_Preferred

Levofloxacin N-oxide is an analytical standard representing a minor metabolite of the antibiotic levofloxacin, supplied for research and analytical applications including method development and metabolite profiling.

  • Provided as an analytical standard for research and method development.
  • Minor metabolite of levofloxacin, useful for metabolite profiling.
  • Chemical formula C18H20FN3O5; molecular weight 377.37 g/mol.
  • High purity (98.28%) suitable for analytical use.
  • Soluble in DMSO at 50 mg/mL; ultrasonic assistance may be required.
  • Store at 4°C and protect from light; in solution keep at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-60523


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.