Learn More
Cayman Chemical Levofloxacin N-oxide, Purity: ≥95%, CAS: 117678-38-3, 25 mg
SDP
Catalog No. 502937812
Encompass_Preferred
An inactive metabolite of levofloxacin; a degradation product of levofloxacin that is formed through exposure to daylight or hydrogen peroxide
Catalog No. 50-293-7812 Supplier Cayman Chemical Supplier No. 2717725 MG
An inactive metabolite of levofloxacin; a degradation product of levofloxacin that is formed through exposure to daylight or hydrogen peroxide