Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levofloxacin Q-Acid 98.0+%, TCI America™
SDP

Catalog No. l02385g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02385G 5 g
L02381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02385G Supplier TCI America Supplier No. L02385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100986-89-8
Molecular Formula C13H9F2NO4
Molecular Weight (g/mol) 281.215
MDL Number MFCD04039905
InChI Key NVKWWNNJFKZNJO-YFKPBYRVSA-N
Synonym levofloxacin q-acid, levofloxacin carboxylic acid, s-9,10-difluoro-3-methyl-7-oxo-3,7-dihydro-2h-1,4 oxazino 2,3,4-ij quinoline-6-carboxylic acid, unii-08gt8fy84e, levofloxacin related compound b, levofloxacin acid, 3s-9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, levofloxacin q-acid,-, s-9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid, levofloxacin related compound b usp
PubChem CID 688333
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 281.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Levofloxacin Q-Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.