Learn More
Medchemexpress LLC Levoglucosan (1,6-anhydro-β-D-glucopyranose) | 498-07-7 | MFCD00063248 | ≥99.0% | 162.14 g/mol | C6H10O5 | 250 MG
SDP
Catalog No. 5000209477
Encompass_Preferred
Levoglucosan (1,6-anhydro-β-D-glucopyranose) is an anhydrosugar produced by glucan pyrolysis and supplied as a high-purity analytical reagent for research use. Typical properties include CAS 498-07-7, molecular formula C6H10O5, and molecular weight 162.14 g/mol.
  • High documented purity (≥99.0%).
  • Available as a 250 mg solid, larger bulk sizes, and 10 mM solutions in DMSO.
  • Soluble in DMSO (100 mg/mL) and compatible with common in vivo formulation systems at ≥2.5 mg/mL.
  • Hygroscopic; use freshly opened DMSO for solution preparation.
  • Stable as powder at -20°C for long-term storage; in solution, store at -80°C for up to 6 months.
Catalog No. 50-002-09477 Supplier Medchemexpress LLC Supplier No. 250MGHYW050145
Levoglucosan (1,6-anhydro-β-D-glucopyranose) is an anhydrosugar produced by glucan pyrolysis and supplied as a high-purity analytical reagent for research use. Typical properties include CAS 498-07-7, molecular formula C6H10O5, and molecular weight 162.14 g/mol.
  • High documented purity (≥99.0%).
  • Available as a 250 mg solid, larger bulk sizes, and 10 mM solutions in DMSO.
  • Soluble in DMSO (100 mg/mL) and compatible with common in vivo formulation systems at ≥2.5 mg/mL.
  • Hygroscopic; use freshly opened DMSO for solution preparation.
  • Stable as powder at -20°C for long-term storage; in solution, store at -80°C for up to 6 months.