Learn More
Medchemexpress LLC Levoglucosenone | 37112-31-5 | 99.4% | 126.11 | 250 MG
SDP
Catalog No. 5000402478
Encompass_Preferred
Levoglucosenone is a biochemical assay reagent with unique chemical properties. It serves as an important intermediate in the production of various chemicals, including pharmaceuticals, fragrances, and flavorings. Due to its ability to dissolve both polar and non-polar compounds, it can also function as a solvent or fuel additive. It is suitable as a biological material or organic compound for life science related research.
  • Suitable as a biological material
  • Functions as an organic compound for life science research
  • Important intermediate in chemical production
  • Can be used as a solvent or fuel additive
Catalog No. 50-004-02478 Supplier Medchemexpress LLC Supplier No. 250MGHY21226
Levoglucosenone is a biochemical assay reagent with unique chemical properties. It serves as an important intermediate in the production of various chemicals, including pharmaceuticals, fragrances, and flavorings. Due to its ability to dissolve both polar and non-polar compounds, it can also function as a solvent or fuel additive. It is suitable as a biological material or organic compound for life science related research.
  • Suitable as a biological material
  • Functions as an organic compound for life science research
  • Important intermediate in chemical production
  • Can be used as a solvent or fuel additive