Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levoleucovorin Calcium | 80433-71-2 | 99.9% | 511.50 | 500 MG
SDP

Catalog No. 5000348763
Encompass_Preferred

Levoleucovorin Calcium, also known as Calcium levofolinate, is the active form of calcium Folinic acid. It exhibits anti-tumor effects and acts as a potentiator for the anti-cancer toxicity of 5-Fluorouracil. This product is intended for research use only.

  • Active form of calcium folinic acid
  • Exhibits anti-tumor effects
  • Potentiates anti-cancer toxicity of 5-Fluorouracil
  • Ships at room temperature (continental US)
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
  • Soluble in water (7.69 mg/mL, requires sonication)
  • Insoluble or slightly soluble in DMSO
  • Appears as a white to yellow solid

Catalog No. 50-003-48763 Supplier Medchemexpress LLC Supplier No. HY13667500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levoleucovorin Calcium, also known as Calcium levofolinate, is the active form of calcium Folinic acid. It exhibits anti-tumor effects and acts as a potentiator for the anti-cancer toxicity of 5-Fluorouracil. This product is intended for research use only.

  • Active form of calcium folinic acid
  • Exhibits anti-tumor effects
  • Potentiates anti-cancer toxicity of 5-Fluorouracil
  • Ships at room temperature (continental US)
  • Store at 4°C, sealed, away from moisture and light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
  • Soluble in water (7.69 mg/mL, requires sonication)
  • Insoluble or slightly soluble in DMSO
  • Appears as a white to yellow solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.