Learn More
Medchemexpress LLC Levomedetomidine hydrochloride | 190000-46-5 | 99.4% | C13H17ClN2 | 10MG

Supplier: Medchemexpress LLC HY135735A10MG
Levomedetomidine hydrochloride is the levorotatory enantiomer of medetomidine, provided as the hydrochloride salt for research applications. It exhibits cardioprotective, sedative, and analgesic effects in preclinical studies and is supplied as a characterized, high-purity research reagent.
- High purity: 99.35%.
- Chemical formula: C13H17ClN2.
- Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
- Recommended storage: solid at 4°C; stock solutions -80°C (up to 6 months) or -20°C (up to 1 month).
- Intended for research use only.
- Available in small package sizes suitable for benchtop studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.