Learn More
Cayman Chemical Levomefolate-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1356019-94-7, 1 mg
SDP
Catalog No. 502992411
Encompass_Preferred
An internal standard for the quantification of levomefolate by GC- or LC-MS
Catalog No. 50-299-2411 Supplier Cayman Chemical Supplier No. 295791 MG
An internal standard for the quantification of levomefolate by GC- or LC-MS