Learn More
Cayman Chemical Levomefolate (calcium salt hydrate), Purity: ≥98%, 1 g
SDP
Catalog No. 502987468
Encompass_Preferred
A biologically active form of folic acid; methyl-group donor involved in the conversion of homocysteine to methionine; important for prevention of neural tube defects
Catalog No. 50-298-7468 Supplier Cayman Chemical Supplier No. 216161 G
A biologically active form of folic acid; methyl-group donor involved in the conversion of homocysteine to methionine; important for prevention of neural tube defects