Learn More
Medchemexpress LLC Levomefolic acid-13C,d3 | 1356019-94-7 | 98.4% | 463.47 | 500 UG
SDP
Catalog No. 5000359772
Encompass_Preferred
Levomefolic acid-13C,d3 is the deuterium labeled Levomefolic acid (HY-14781). It is an orally active, brain-penetrant natural active form of folic acid and a widely used folic acid food supplement. This compound can be used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated for drug development quantitation. Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Orally active, brain-penetrant natural active form of folic acid
  • Widely used folic acid food supplement
  • Used as a tracer and internal standard for quantitative analysis
  • Stable heavy isotopes incorporated for drug development quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-59772 Supplier Medchemexpress LLC Supplier No. HY14781S1500UG
Levomefolic acid-13C,d3 is the deuterium labeled Levomefolic acid (HY-14781). It is an orally active, brain-penetrant natural active form of folic acid and a widely used folic acid food supplement. This compound can be used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated for drug development quantitation. Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Orally active, brain-penetrant natural active form of folic acid
  • Widely used folic acid food supplement
  • Used as a tracer and internal standard for quantitative analysis
  • Stable heavy isotopes incorporated for drug development quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles