Learn More
Apexbio Technology LLC LEVOMEFOLIC-ACID-50MG
SDP
Catalog No. 5000567533
Encompass_Preferred
Levomefolic acid (CAS No 31690-09-2) is a prominent compound in biomedical research known for its role as the biologically active form of folate Originating as a metabolite of dihydrofolate it participates in pathways essential for DNA synthesis and repair primarily targeting the one-carbon cycle Its mechanism involves facilitating the conversion of homocysteine to methionine impacting cellular methylation processes In vitro levomefolic acid exhibits activity in the nanomolar to low micromolar range crucial for studies on cell proliferation and differentiation It is frequently utilized in research involving various cell models and animal studies including those focusing on neurodevelopment and cancer The compound is also integral to drug screening assays where folate metabolism is implicated Experimental concentrations typically depend on the specific research design adjusting based on the objectives and conditions of each study
Catalog No. 50-005-67533 Supplier Apexbio Technology LLC Supplier No. C865050
Add to Cart
Add to Cart
Levomefolic acid (CAS No 31690-09-2) is a prominent compound in biomedical research known for its role as the biologically active form of folate Originating as a metabolite of dihydrofolate it participates in pathways essential for DNA synthesis and repair primarily targeting the one-carbon cycle Its mechanism involves facilitating the conversion of homocysteine to methionine impacting cellular methylation processes In vitro levomefolic acid exhibits activity in the nanomolar to low micromolar range crucial for studies on cell proliferation and differentiation It is frequently utilized in research involving various cell models and animal studies including those focusing on neurodevelopment and cancer The compound is also integral to drug screening assays where folate metabolism is implicated Experimental concentrations typically depend on the specific research design adjusting based on the objectives and conditions of each study