Learn More
Selleck Chemical LLC Levomefolic acid
SDP
Catalog No. 5000002426
Encompass_Preferred
Levomefolic acid (5-MTHF LMSR Metafolin Nutrifolin) is a natural active form of folic acid used at the cellular level for DNA reproduction the cysteine cycle and the regulation of homocysteine among other functions
Catalog No. 50-000-02426 Supplier Selleck Chemical LLC Supplier No. S69095MG
Add to Cart
Add to Cart
Levomefolic acid (5-MTHF LMSR Metafolin Nutrifolin) is a natural active form of folic acid used at the cellular level for DNA reproduction the cysteine cycle and the regulation of homocysteine among other functions