Learn More
Medchemexpress LLC Levomilnacipran hydrochloride | 175131-60-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0168B10MM/1ML
Encompass_Preferred
Levomilnacipran hydrochloride, also known as (1S,2R)-Milnacipran hydrochloride or F-2695 hydrochloride, is the enantiomer of Milnacipran. It acts as a strong substrate of P-gp, enabling it to cross the blood-brain barrier. This compound is a serotonin and norepinephrine reuptake inhibitor, demonstrating antidepressant and anxiolytic activities, and is used in depression research.
  • Acts as a strong substrate of P-gp
  • Enables crossing the blood-brain barrier
  • Serotonin and norepinephrine reuptake inhibitor
  • Demonstrates antidepressant and anxiolytic activities
  • Used in depression research
  • Has IC50 values of 10.5 nM and 19.0 nM
  • Has Ki values of 92.2 nM and 1.2 nM
  • Purity of 99.95%
Catalog No. 50-004-06289