Learn More
Medchemexpress LLC Levomilnacipran hydrochloride | 175131-60-9 | 99.95% | 100 MG
SDP
Catalog No. 5000405990
Encompass_Preferred
Levomilnacipran hydrochloride is the enantiomer of Milnacipran and a strong P-gp substrate capable of crossing the blood-brain barrier. It acts as a serotonin and norepinephrine reuptake inhibitor, demonstrating antidepressant and anxiolytic activities, making it suitable for depression research.
  • Strong P-gp substrate, able to cross the blood-brain barrier.
  • Serotonin and norepinephrine reuptake inhibitor.
  • Exhibits antidepressant and anxiolytic activities.
  • Used for research on depression.
  • Ameliorates depression-like behaviors in rat models.
  • Improves neuronal synaptic deficits and morphological changes.
  • Upregulates synaptic-related proteins like PSD-95 and Syn.
  • Activates the BDNF/TrkB mediated PI3K/Akt/mTOR signaling pathway.
  • Suppresses neuroinflammation.
Catalog No. 50-004-05990 Supplier Medchemexpress LLC Supplier No. HYB0168B100MG
Levomilnacipran hydrochloride is the enantiomer of Milnacipran and a strong P-gp substrate capable of crossing the blood-brain barrier. It acts as a serotonin and norepinephrine reuptake inhibitor, demonstrating antidepressant and anxiolytic activities, making it suitable for depression research.
  • Strong P-gp substrate, able to cross the blood-brain barrier.
  • Serotonin and norepinephrine reuptake inhibitor.
  • Exhibits antidepressant and anxiolytic activities.
  • Used for research on depression.
  • Ameliorates depression-like behaviors in rat models.
  • Improves neuronal synaptic deficits and morphological changes.
  • Upregulates synaptic-related proteins like PSD-95 and Syn.
  • Activates the BDNF/TrkB mediated PI3K/Akt/mTOR signaling pathway.
  • Suppresses neuroinflammation.