Learn More
Medchemexpress LLC Levomilnacipran (hydrochloride) | 175131-60-9 | 99.95% | 25 MG
SDP
Catalog No. 5000450560
Encompass_Preferred
Levomilnacipran hydrochloride is the enantiomer of Milnacipran and a potent serotonin and norepinephrine reuptake inhibitor. It is a strong substrate of P-gp that can cross the blood-brain barrier, exhibiting antidepressant and anxiolytic activities. It is suitable for research on depression.
  • Serotonin and norepinephrine reuptake inhibitor
  • Exhibits antidepressant and anxiolytic activities
  • Crosses the blood-brain barrier
  • Used for research of depression
Catalog No. 50-004-50560 Supplier Medchemexpress LLC Supplier No. 25MGHYB0168B
Levomilnacipran hydrochloride is the enantiomer of Milnacipran and a potent serotonin and norepinephrine reuptake inhibitor. It is a strong substrate of P-gp that can cross the blood-brain barrier, exhibiting antidepressant and anxiolytic activities. It is suitable for research on depression.
  • Serotonin and norepinephrine reuptake inhibitor
  • Exhibits antidepressant and anxiolytic activities
  • Crosses the blood-brain barrier
  • Used for research of depression