Learn More
Apexbio Technology LLC Levonorgestrel 797-63-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19605.1
Encompass_Preferred
Levonorgestrel is a synthetic progesterone analog structurally categorized as a progestin exhibiting agonistic activity primarily through progesterone receptor (PR) binding and also showing interaction with estrogen receptors (ER) Upon receptor binding levonorgestrel modulates transcriptional activity influencing downstream gene expression related to reproductive processes This molecular interaction reduces luteinizing hormone levels inhibits ovulation and induces alterations in uterine lining and cervical mucus thus contributing to contraceptive activity Biomedically levonorgestrel is widely utilized in research for studying progesterone receptor signaling pathways hormonal regulation mechanisms and contraceptive drug development
Catalog No. 50-000-62018