Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Levonorgestrel (LNG) Competitive ELISA Kit

Catalog No. EIALNGCX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIALNGCX10 10 x 96 Tests
EIALNGC 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIALNGCX10 Supplier Invitrogen™ Supplier No. EIALNGCX10
Add to Cart
Add to Cart
Request a Quote

ELISA

The Levonorgestrel (LNG) ELISA quantitates LNG in serum,plasma,urine,dried fecal extracts,saliva,milk,or cell culture medium.

Principle of the method

The Levonorgestrel (LNG) Competitive ELISA research-use-only kit is designed to quantitatively measure levonorgestrel present in serum,plasma,saliva,urine,dried fecal samples,milk,and tissue culture media samples. This kit measures total LNG in extracted serum,plasma,milk and fecal samples. A levonorgestrel stock solution is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. A levonorgestrel-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a rabbit polyclonal antibody to levonorgestrel to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound levonorgestrel-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Levonorgestrel (4-Estren-17alpha-Ethynyl-18-homo-17beta-ol-3-one (LNG) is a synthetic steroid analogue.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 6.25-200 pg/mL
Assay Sensitivity 2.2 pg/mL
Description Levonorgestrel (LNG) Competitive ELISA Kit
Assay Multiplex Assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Milk, Plasma, Saliva, Serum, Supernatant, Urine, Water
Sufficient For 10 x 96 Assays
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV 9.1%
Intraassay CV 9%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases C21H28O2
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 100 μL, Milk 100 μL, Plasma 100 μL, Saliva 50 μL, Serum 100 μL, Supernatant 100 μL, Urine 25 μL, Water 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Levonorgestrel
Test Time 1 hr 20 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.