Learn More
Arbor Assays Levonorgestrel (LNG) ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K058H1
Encompass_Preferred
The DetectX Levonorgestrel (LNG) ELISA Kits are designed to quantitatively measure Levonorgestrel present in serum, plasma, saliva, urine, dried fecal samples, milk, and tissue culture media samples. 
Catalog No. 50-227-8484