Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levonorgestrel (standard) | 797-63-7 | 99.9% | 100 MG
SDP

Catalog No. 5000406187
Encompass_Preferred

Levonorgestrel (Standard) is an analytical standard of Levonorgestrel, intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments, such as HPLC, GC, and MS. With a molecular weight of 312.45 and a chemical formula of C₂₁H₂₈O₂, it appears as a solid, white to off-white in color.

  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS.
  • Molecular weight of 312.45.
  • Chemical formula is C₂₁H₂₈O₂.
  • Appears as a solid, white to off-white in color.

Catalog No. 50-004-06187 Supplier Medchemexpress LLC Supplier No. HYB0257R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levonorgestrel (Standard) is an analytical standard of Levonorgestrel, intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments, such as HPLC, GC, and MS. With a molecular weight of 312.45 and a chemical formula of C₂₁H₂₈O₂, it appears as a solid, white to off-white in color.

  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS.
  • Molecular weight of 312.45.
  • Chemical formula is C₂₁H₂₈O₂.
  • Appears as a solid, white to off-white in color.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.