Learn More
Medchemexpress LLC Levonorgestrel (Standard) | 797-63-7 | 99.9% | 25 MG
SDP
Catalog No. 5000450539
Encompass_Preferred
Levonorgestrel (Standard) is the analytical standard of Levonorgestrel, intended for research and analytical applications. This orally active inhibitor of progesterone has anticancer activity and can induce apoptosis. It is utilized in qualitative, quantitative, and methodological research experiments, as well as in the study of osteoporosis and uterine leiomyoma.
  • Analytical standard for research and analytical applications
  • Commonly used in HPLC, GC, and MS experiments
  • Functions as an orally active inhibitor of progesterone
  • Exhibits anticancer activity and induces apoptosis
  • Can be used as a contraceptive and in combination with other medications
  • Inhibits cell proliferation and promotes apoptosis in uterine leiomyoma cells
  • Can inhibit bone resorption and increase bone mineral content in in vivo studies
Catalog No. 50-004-50539 Supplier Medchemexpress LLC Supplier No. 25MGHYB0257R
Levonorgestrel (Standard) is the analytical standard of Levonorgestrel, intended for research and analytical applications. This orally active inhibitor of progesterone has anticancer activity and can induce apoptosis. It is utilized in qualitative, quantitative, and methodological research experiments, as well as in the study of osteoporosis and uterine leiomyoma.
  • Analytical standard for research and analytical applications
  • Commonly used in HPLC, GC, and MS experiments
  • Functions as an orally active inhibitor of progesterone
  • Exhibits anticancer activity and induces apoptosis
  • Can be used as a contraceptive and in combination with other medications
  • Inhibits cell proliferation and promotes apoptosis in uterine leiomyoma cells
  • Can inhibit bone resorption and increase bone mineral content in in vivo studies