Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Levonorgestrel (Standard) | 797-63-7 | 99.94% | 50 MG
SDP

Catalog No. 5000407366
Encompass_Preferred

Levonorgestrel (Standard) is the analytical standard of Levonorgestrel, intended for research and analytical applications. This product appears as a white to off-white solid. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Used in qualitative, quantitative, and methodological research experiments
  • Appearance: white to off-white solid
  • Storage (powder): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years; -20°C for 1 year

Catalog No. 50-004-07366 Supplier Medchemexpress LLC Supplier No. HYB0257R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levonorgestrel (Standard) is the analytical standard of Levonorgestrel, intended for research and analytical applications. This product appears as a white to off-white solid. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.

  • Used in qualitative, quantitative, and methodological research experiments
  • Appearance: white to off-white solid
  • Storage (powder): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years; -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.