Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levonorgestrel, Tocris Bioscience™
SDP

Catalog No. 483350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 48-335-0 Supplier R&D Systems Supplier No. 4833/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic progesterone analog

Synthetic progesterone analog; binds to the progesterone receptor (relative binding affinities are < 0.02, 7.5, 17, 58 and 323 % for estrogen receptors, glucocorticoid receptors, mineralocorticoid receptors, androgen receptors and progesterone receptors respectively).

Chemical Identifiers

CAS 797-63-7
Molecular Formula C21H28O2
Molecular Weight (g/mol) 312.453
InChI Key WWYNJERNGUHSAO-XUDSTZEESA-N
Synonym levonorgestrel, norgestrel, d-norgestrel, --norgestrel, levonova, microval, postinor, mirena, plan b, jadelle
PubChem CID 13109
ChEBI CHEBI:6443
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CCC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34

Specifications

Quantity 50 mg
Formula Weight 312.45
Percent Purity >98%
Chemical Name or Material (17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.