Learn More
Medchemexpress LLC Levophacetoperane hydrochloride | 23257-56-9 | 98.0% | 269.77 g/mol | C14H20ClNO2 | 1 ML
SDP
Catalog No. 5000249075
Encompass_Preferred
Levophacetoperane hydrochloride is a research chemical that competitively inhibits norepinephrine and dopamine uptake in vitro. It is supplied for laboratory use in both solid form and as a ready-to-use 10 mM solution in DMSO, and is intended for non-clinical research applications.
  • Inhibits norepinephrine and dopamine uptake in vitro.
  • Available as solid and 10 mM solution in DMSO for flexible use.
  • High purity (98.0%) suitable for pharmacology studies.
  • Soluble in DMSO (≈61.8 mM); ultrasonic assistance may be needed.
  • Store sealed away from moisture; in solvent: -80 °C (6 months), -20 °C (1 month).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-49075 Supplier Medchemexpress LLC Supplier No. HY10163110MM 1ML
Levophacetoperane hydrochloride is a research chemical that competitively inhibits norepinephrine and dopamine uptake in vitro. It is supplied for laboratory use in both solid form and as a ready-to-use 10 mM solution in DMSO, and is intended for non-clinical research applications.
  • Inhibits norepinephrine and dopamine uptake in vitro.
  • Available as solid and 10 mM solution in DMSO for flexible use.
  • High purity (98.0%) suitable for pharmacology studies.
  • Soluble in DMSO (≈61.8 mM); ultrasonic assistance may be needed.
  • Store sealed away from moisture; in solvent: -80 °C (6 months), -20 °C (1 month).
  • For research use only; not for human or clinical use.