Learn More
TARGETMOL CHEMICALS INC LEVOSEMOTIADIL 5MG
SDP
Catalog No. 502580741
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Levosemotiadil(SA 3212) is a novel calcium antagonist and a very potent inhibitor of low-density lipoprotein oxidation.Levosemotiadil may be used to prevent lethal cardiac arrhythmias. purity: 99%
Catalog No. 50-258-0741 Supplier TARGETMOL CHEMICALS INC Supplier No. T257015MG
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Levosemotiadil(SA 3212) is a novel calcium antagonist and a very potent inhibitor of low-density lipoprotein oxidation.Levosemotiadil may be used to prevent lethal cardiac arrhythmias. purity: 99%