Learn More
Apexbio Technology LLC Levosimendan 141505-33-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061742
Encompass_Preferred
Levosimendan (CAS 141505-33-1) is a small molecule calcium sensitizer that exerts its pharmacological action by binding selectively and in a calcium-dependent manner to cardiac troponin C (cTnC) Through this interaction levosimendan facilitates cardiac muscle contraction without significantly increasing intracellular calcium concentrations Research indicates utility in heart failure treatment where improved myocardial contractility is therapeutically desirable Currently levosimendan is evaluated extensively in Phase 4 clinical studies to explore its efficacy and therapeutic profile
Catalog No. 50-000-61742 Supplier Apexbio Technology LLC Supplier No. B19615.1
Levosimendan (CAS 141505-33-1) is a small molecule calcium sensitizer that exerts its pharmacological action by binding selectively and in a calcium-dependent manner to cardiac troponin C (cTnC) Through this interaction levosimendan facilitates cardiac muscle contraction without significantly increasing intracellular calcium concentrations Research indicates utility in heart failure treatment where improved myocardial contractility is therapeutically desirable Currently levosimendan is evaluated extensively in Phase 4 clinical studies to explore its efficacy and therapeutic profile