Learn More
Medchemexpress LLC Levosulpiride-d3 | 124020-27-5 | 99.8% | 1 MG
SDP
Catalog No. 5000414187
Encompass_Preferred
Levosulpiride-d3 is the deuterium labeled Levosulpiride, an (S)-enantiomer of sulpiride that acts as a D2 receptor antagonist and atypical antipsychotic agent of the benzamide class. This compound is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-004-14187 Supplier Medchemexpress LLC Supplier No. HYB1059S1MG
Levosulpiride-d3 is the deuterium labeled Levosulpiride, an (S)-enantiomer of sulpiride that acts as a D2 receptor antagonist and atypical antipsychotic agent of the benzamide class. This compound is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.