Learn More
Medchemexpress LLC Levosulpiride-d3 | 124020-27-5 | 99.8% | 5 MG
SDP
Catalog No. 5000413773
Encompass_Preferred
Levosulpiride-d3 is the deuterium labeled Levosulpiride. Levosulpiride (RV-12309) is the (S)-enantiomer of sulpiride, which is a D2 receptor antagonist, an atypical antipsychotic agent of the benzamide class.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • D3 receptor antagonist
  • Used in neurological, eye or ear, metabolic or endocrine diseases
Catalog No. 50-004-13773 Supplier Medchemexpress LLC Supplier No. HYB1059S5MG
Levosulpiride-d3 is the deuterium labeled Levosulpiride. Levosulpiride (RV-12309) is the (S)-enantiomer of sulpiride, which is a D2 receptor antagonist, an atypical antipsychotic agent of the benzamide class.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • D3 receptor antagonist
  • Used in neurological, eye or ear, metabolic or endocrine diseases