Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Levothyroxine Sodium, USP
SDP

Catalog No. 14186163 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-163 500 mg
14-185-479 25 g
14-185-535 5 g
14-186-242 100 g
14-186-241 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-186-163 Supplier Medisca Supplier No. 165700
Add to Cart
Add to Cart

Levothyroxine Sodium USP is a light yellow to buff-colored, odorless, hygroscopic powder. It is stable in dry air but may assume a slight pink color upon exposure to light.

Levothyroxine Sodium USP is a light yellow to buff-colored, odorless, hygroscopic powder. It is stable in dry air but may assume a slight pink color upon exposure to light.

Alternative name(s): Thyroxine sodium, Levothyroxine sodium monohydrate, L-Thyroxine sodium xhydrate

Specifications

Chemical Name or Material Levothyroxine Sodium
Packaging Bottle, Amber Glass
CAS 25416-65-3
Type Drug Standard
Molecular Weight (g/mol) 798.86
Quantity 500 mg
Percent Purity 97 to 103 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.