Learn More
Medchemexpress LLC HY-Y0839 100mg Medchemexpress, Levulinic acid CAS:123-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0839100MG
Encompass_Preferred
Medchemexpress, HY-Y0839 100mg Levulinic acid CAS:123-76-2 Levulinic acid is a precursor for the synthesis of biofuels, such as ethyl levulinate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0766