Learn More
Medchemexpress LLC Levulinic acid | 123-76-2 | MFCD00002796 | 100.0% | 116.12 g/mol | C5H8O3 | 100g
SDP
Catalog No. 5000268616
Encompass_Preferred
Levulinic acid is a precursor for the synthesis of biofuels such as ethyl levulinate Levulinic acid is also a 5-keto-pentanoic acid Levulinic acid can be utilized by the cells as a cosubstrate for biopolymer synthesis Levulinic acid can be used in antibacterial research[1][2][3]
Catalog No. 50-002-68616 Supplier Medchemexpress LLC Supplier No. HYY0839100G
Levulinic acid is a precursor for the synthesis of biofuels such as ethyl levulinate Levulinic acid is also a 5-keto-pentanoic acid Levulinic acid can be utilized by the cells as a cosubstrate for biopolymer synthesis Levulinic acid can be used in antibacterial research[1][2][3]