Learn More
Andwin Scientific LEVY DOUBLE COUNT CHAMBER
SDP
Catalog No. NC9947675
Encompass_Preferred
Leavy Counting Chamber, Hemacytometer for accurate red and white blood cell counts. Improved double Neubauer rulings with dark lines on a clear background. Cell depth: 0.1mm 1-piece construction. Dimensions: 75L x 32W x 4.5mm D
Catalog No. NC9947675 Supplier Andwin Scientific Supplier No. 15170208
Leavy Counting Chamber, Hemacytometer for accurate red and white blood cell counts. Improved double Neubauer rulings with dark lines on a clear background. Cell depth: 0.1mm 1-piece construction. Dimensions: 75L x 32W x 4.5mm D