Learn More
MilliporeSigma™ Supelco™ Lewatit™ VP OC 1065
Catalog No. 111005021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-5021 100 g
1 options
Catalog No. 11-100-5021 Supplier MilliporeSigma™ Supelco™ Supplier No. 94136100GF
Add to Cart
Add to Cart
Adsorber resin functionalised with benzyl amine groups
Weakly basic anion exchange resin
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g
Product Line Lewatit
Capacity (Metric) 1.4 meq/mL
MDL Number MFCD05865453
Lewatit is a registered trademark of Lanxess Deutschland GmbH