Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM21412P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: A70-A/A9. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Lung, Colon, Bladder or Hepatocellular Carcinoma. Cellular Staining: Cell membrane, cytoplasmic and extracellular. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Live Ls174T cells (human colon carcinoma cell line). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5450