Learn More
MSC Stanco Indura Coverall, 9 oz., 11.5 cal/cm² Arc Rating, 1 ea
Catalog No. 5000686782
Encompass
These Indura® coveralls are designed for protection against arc flash and flash fire hazards. Constructed from 9 oz/yd² flame-resistant cotton fabric, this garment offers a hazard risk category of CAT 2 and an arc rating of 11.5 cal/cm².
  • Features a two-way concealed brass zipper for ease of use
  • Includes an elasticized waistband and full action back for improved mobility
  • Provides ample storage with two front, two rear, two chest, and two pass-through pockets
  • Equipped with a dedicated tool pocket on the leg
Catalog No. 50-006-86782 Supplier MSC Supplier No. 03747599
These Indura® coveralls are designed for protection against arc flash and flash fire hazards. Constructed from 9 oz/yd² flame-resistant cotton fabric, this garment offers a hazard risk category of CAT 2 and an arc rating of 11.5 cal/cm².
  • Features a two-way concealed brass zipper for ease of use
  • Includes an elasticized waistband and full action back for improved mobility
  • Provides ample storage with two front, two rear, two chest, and two pass-through pockets
  • Equipped with a dedicated tool pocket on the leg