Learn More
Fine Science Tools Lexer-Baby Scissors, Stainless Steel, Curved, 10 cm
SDP
Catalog No. NC9818585
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC9818585 Supplier Fine Science Tools Supplier No. 1407910
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 26mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No