Learn More
TARGETMOL CHEMICALS INC LEXIBULIN 10MG
SDP
Catalog No. 502581032
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lexibulin (CYT-997)(CYT-997) is a potent tubulin polymerization inhibitor (IC50 10-100 nM in Y cell lines). Lexibulin blocks the formation of the mitotic spindle and leading to cell cycle arrest at the G2/M phase; this may result in disruption of the tumor vasculature and tumor blood flow and tumor cell death. purity: 99%
Catalog No. 50-258-1032 Supplier TARGETMOL CHEMICALS INC Supplier No. T209010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lexibulin (CYT-997)(CYT-997) is a potent tubulin polymerization inhibitor (IC50 10-100 nM in Y cell lines). Lexibulin blocks the formation of the mitotic spindle and leading to cell cycle arrest at the G2/M phase; this may result in disruption of the tumor vasculature and tumor blood flow and tumor cell death. purity: 99%