Learn More
Cayman Chemical LexIthromycIn 25mg
SDP
Catalog No. 502987148
Encompass_Preferred
A macrolide antibiotic; a semisynthetic erythromycin derivative; improved pH stability and absorption over erythromycin; prevents protein synthesis; binds ribosome polypeptide exit tunnel
Catalog No. 50-298-7148 Supplier Cayman Chemical Supplier No. 2200225 MG
A macrolide antibiotic; a semisynthetic erythromycin derivative; improved pH stability and absorption over erythromycin; prevents protein synthesis; binds ribosome polypeptide exit tunnel