Learn More
Ideal Vacuum Products LLC LEYBOLD RE 8-16 REPLACEMENT FI
SDP
Catalog No. NC1832030
Encompass
Leybold RE 8-16 Replacement Filter for Leybold SE 8-16 Smoke Eliminator
Catalog No. NC1832030 Supplier Ideal Vacuum Products LLC Supplier No. P102973
Leybold RE 8-16 Replacement Filter for Leybold SE 8-16 Smoke Eliminator