Learn More
TARGETMOL CHEMICALS INC LF3 25MG
SDP
Catalog No. 502579195
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LF3 is a specific inhibitor of canonical Wnt signaling by disrupting the interaction between (beta)-catenin and TCF4 (IC50 < 2 uM)purity: 98%
Catalog No. 50-257-9195 Supplier TARGETMOL CHEMICALS INC Supplier No. T739925MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LF3 is a specific inhibitor of canonical Wnt signaling by disrupting the interaction between (beta)-catenin and TCF4 (IC50 < 2 uM)purity: 98%