Learn More
Medchemexpress LLC HY-101486 5mg Medchemexpress, LF3 CAS:664969-54-4 Purity:>98%
SDP
Catalog No. 501932147
Encompass_Preferred
Medchemexpress, HY-101486 5mg LF3 CAS:664969-54-4 LF3 is an antagonist of the β-Catenin/TCF4 interaction with antitumor activity; has an IC 50 of 1.65 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2147 Supplier Medchemexpress LLC Supplier No. HY1014865MG
Medchemexpress, HY-101486 5mg LF3 CAS:664969-54-4 LF3 is an antagonist of the β-Catenin/TCF4 interaction with antitumor activity; has an IC 50 of 1.65 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.