Learn More
Sino Biological L-FABP Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12353T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human L-FABP (rh L-FABP; Catalog#12353-H07E; NP_001434.1; Ser2-Ile127). L-FABP specific IgG was purified by Human L-FABP affinity chromatography.
Catalog No. 50-224-6386