Learn More
Medchemexpress LLC Lfhp-1c | 2102347-47-5 | 98.0% | 873.13 | 100 MG
SDP
Catalog No. 5000350366
Encompass_Preferred
LFHP-1c is a PGAM5 inhibitor with neuroprotective activity in brain ischemic stroke. It protects blood-brain barrier integrity from ischemia-induced injury. LFHP-1c binds to endothelial PGAM5 to inhibit the activity of PGAM5 phosphatase and its interaction with NRF2. It exhibits in vitro and in vivo protection.
  • PGAM5 inhibitor
  • Offers neuroprotective activity in brain ischemic stroke
  • Protects blood-brain barrier integrity from ischemia-induced injury
  • Binds to endothelial PGAM5 to inhibit the activity of PGAM5 phosphatase
  • Inhibits the interaction of PGAM5 with NRF2
  • Exhibits in vitro and in vivo protection
Catalog No. 50-003-50366 Supplier Medchemexpress LLC Supplier No. HY139598100MG
LFHP-1c is a PGAM5 inhibitor with neuroprotective activity in brain ischemic stroke. It protects blood-brain barrier integrity from ischemia-induced injury. LFHP-1c binds to endothelial PGAM5 to inhibit the activity of PGAM5 phosphatase and its interaction with NRF2. It exhibits in vitro and in vivo protection.
  • PGAM5 inhibitor
  • Offers neuroprotective activity in brain ischemic stroke
  • Protects blood-brain barrier integrity from ischemia-induced injury
  • Binds to endothelial PGAM5 to inhibit the activity of PGAM5 phosphatase
  • Inhibits the interaction of PGAM5 with NRF2
  • Exhibits in vitro and in vivo protection