Learn More
Apexbio Technology LLC LFM-A13 244240-24-2 200mg
SDP
Catalog No. 5000095326
Encompass_Preferred
LFM-A13 (CAS 244240-24-2) is a small-molecule inhibitor targeting Bruton s tyrosine kinase (BTK) a cytoplasmic enzyme implicated in the signaling pathways regulating the growth and differentiation of B-lineage lymphoid cells LFM-A13 demonstrates high selectivity for BTK exhibiting an IC50 value of 17 2 M in kinase assays and shows minimal activity against other protein tyrosine kinases even at elevated concentrations In preclinical models LFM-A13 enhances the efficacy of standard chemotherapeutic regimens and increases survival rates in mice challenged with BCL-1 leukemia cells Currently LFM-A13 is under investigation in preclinical research settings
Catalog No. 50-000-95326 Supplier Apexbio Technology LLC Supplier No. B5952200
LFM-A13 (CAS 244240-24-2) is a small-molecule inhibitor targeting Bruton s tyrosine kinase (BTK) a cytoplasmic enzyme implicated in the signaling pathways regulating the growth and differentiation of B-lineage lymphoid cells LFM-A13 demonstrates high selectivity for BTK exhibiting an IC50 value of 17 2 M in kinase assays and shows minimal activity against other protein tyrosine kinases even at elevated concentrations In preclinical models LFM-A13 enhances the efficacy of standard chemotherapeutic regimens and increases survival rates in mice challenged with BCL-1 leukemia cells Currently LFM-A13 is under investigation in preclinical research settings