Learn More
TARGETMOL CHEMICALS INC LFM-A13 25MG
SDP
Catalog No. 502579483
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LFM-A13(IC50=2.5 uM) a specific Bruton's tyrosine kinase (BTK) is more than 100-fold specificity than other protein kinases such as JAK1 JAK2 HCK EGFR and IRK. purity: 99%
Catalog No. 50-257-9483 Supplier TARGETMOL CHEMICALS INC Supplier No. T621725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LFM-A13(IC50=2.5 uM) a specific Bruton's tyrosine kinase (BTK) is more than 100-fold specificity than other protein kinases such as JAK1 JAK2 HCK EGFR and IRK. purity: 99%