Learn More
Medchemexpress LLC HY-18009 5mg Medchemexpress, LFM-A13 CAS:244240-24-2 Purity:>98%
SDP
Catalog No. 502082010
Encompass_Preferred
Medchemexpress, HY-18009 5mg LFM-A13 CAS:244240-24-2 LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM; LFM-A13 shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR and IRK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2010 Supplier Medchemexpress LLC Supplier No. HY180095MG
Medchemexpress, HY-18009 5mg LFM-A13 CAS:244240-24-2 LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM; LFM-A13 shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR and IRK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.