Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Lg Kimwipe Holder - Double
SDP

Catalog No. 01259206
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-206 Magnetic Mount
01-259-185 VHB Tape
01-259-151 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-206 Supplier Poltex Supplier No. LGLABDBLM
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

Fits two large boxes of laboratory wipes such as Kimwipes. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable and cleanable PETG construction.

  • Fits two large boxes of laboratory wipes (Kimwipes or other).
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Product Type Kim Wipe Dispenser
Color Clear
Dimensions (L x W x H) 4.1 x 11 x 13.7 in.
Holds Wipes, Labwipes
Material PETG
Mounting Magnetic Mount
No. of Compartments 2
Product Style Side Loading
Capacity 2 Box
Edges Rounded
Formulation PETG
For Use With (Application) Holds Two Large Boxes of Laboratory Wipes
Includes Magnets for Mounting
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.