Learn More
Bellco Glass, Inc. Large Nylon Mesh,23x23cm-20/cs
SDP
Catalog No. 501946505
Encompass_Preferred
Used for auto blot hybridization ovens Pore size: 125 microns open area; 39%--Mesh count: (per inch); 127--Weave type: 1/1 plain fabric--Thickness: 120 microns--Weight: 1.5 Ounces per sq yd Size: 23x23cm. MADE IN THE USA
Catalog No. 50-194-6505 Supplier Bellco Glass, Inc. Supplier No. 791002024
Used for auto blot hybridization ovens Pore size: 125 microns open area; 39%--Mesh count: (per inch); 127--Weave type: 1/1 plain fabric--Thickness: 120 microns--Weight: 1.5 Ounces per sq yd Size: 23x23cm. MADE IN THE USA