Learn More
MSC Vikan Hand Brush, 13 in Length, Medium, Red, 1 Each
Catalog No. 502869495
Encompass
This hand brush is specifically designed for effective removal of flour and fine particles from various surfaces like conveyor belts, food preparation areas, tables, and equipment. Its robust construction makes it ideal for use in hygiene and food safety critical environments.
  • Purpose-built for environments where hygiene and food safety are critical
  • Medium bristles allow for both wet and dry cleaning applications
  • Ergonomic design enhances comfort and reduces worker strain
  • Available in multiple colors to support hygienic zoning plans and 5S lean programs
  • Durable construction for long-lasting performance
  • Easy to clean and maintain, aiding hygiene control
  • Features a drop-shaped hanging hole to prevent liquid pooling and facilitate easy storage
Catalog No. 50-286-9495 Supplier MSC Supplier No. 47162839
This hand brush is specifically designed for effective removal of flour and fine particles from various surfaces like conveyor belts, food preparation areas, tables, and equipment. Its robust construction makes it ideal for use in hygiene and food safety critical environments.
  • Purpose-built for environments where hygiene and food safety are critical
  • Medium bristles allow for both wet and dry cleaning applications
  • Ergonomic design enhances comfort and reduces worker strain
  • Available in multiple colors to support hygienic zoning plans and 5S lean programs
  • Durable construction for long-lasting performance
  • Easy to clean and maintain, aiding hygiene control
  • Features a drop-shaped hanging hole to prevent liquid pooling and facilitate easy storage