Learn More
MSC Vikan Broom, Fine Particle, 16.1 in, Stiff Bristle, Pink
Catalog No. 502837505
Encompass
This Vikan broom is engineered with a durable plastic block and features a European threaded handle connection for secure attachment. It is designed for efficient cleaning of fine particles, offering a 16.1 inch brush width and stiff polyester bristles.
  • Plastic block construction
  • European threaded handle connection
  • Fine particle brush type
  • 16.1 inch brush width
  • Polyester bristle material
  • 2 inch bristle length
  • Pink bristle color
  • Stiff bristle stiffness
Catalog No. 50-283-7505 Supplier MSC Supplier No. 47164678
This Vikan broom is engineered with a durable plastic block and features a European threaded handle connection for secure attachment. It is designed for efficient cleaning of fine particles, offering a 16.1 inch brush width and stiff polyester bristles.
  • Plastic block construction
  • European threaded handle connection
  • Fine particle brush type
  • 16.1 inch brush width
  • Polyester bristle material
  • 2 inch bristle length
  • Pink bristle color
  • Stiff bristle stiffness