Learn More
Grainger Qmark SmartSeries Recessed Wall-Mount Heater, 4000 W, 277 V, 1 ea
Catalog No. 502768876
Encompass_Preferred
This recessed electric wall-mount heater provides high-output climate control through fan-forced heating, utilizing proportional wattage adjustment for energy efficiency. Designed for integration with building management systems, the unit features an LED touchscreen programmable thermostat with 5/2-day scheduling and four periods per day, ensuring temperature regulation within 1.5°F.
  • Compatible with building management systems for centralized operation
  • Maintains set temperature within 1.5°F
  • Features non-glowing heating elements and a totally enclosed fan motor
  • Includes a power on/off disconnect switch for maintenance safety
  • Built-in fan delay control optimizes heat distribution
  • Three-piece construction for recessed installation between standard wall studs
  • Includes remote control for temperature and on/off adjustments
  • White powder-coated finish
Catalog No. 50-276-8876 Supplier Grainger Supplier No. 13R110
This recessed electric wall-mount heater provides high-output climate control through fan-forced heating, utilizing proportional wattage adjustment for energy efficiency. Designed for integration with building management systems, the unit features an LED touchscreen programmable thermostat with 5/2-day scheduling and four periods per day, ensuring temperature regulation within 1.5°F.
  • Compatible with building management systems for centralized operation
  • Maintains set temperature within 1.5°F
  • Features non-glowing heating elements and a totally enclosed fan motor
  • Includes a power on/off disconnect switch for maintenance safety
  • Built-in fan delay control optimizes heat distribution
  • Three-piece construction for recessed installation between standard wall studs
  • Includes remote control for temperature and on/off adjustments
  • White powder-coated finish